missing translation for 'onlineSavingsMsg'
Learn More

Iron boride, 98%

Catalog No. p-7076506
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8814622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA8814622 Supplier Thermo Scientific Chemicals Supplier No. 08814622
Only null left

Used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. | CAS: 12006-84-7 | BFe | 66.66 g/mol

Iron boride is used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. Being inexpensive as well as having excellent mechanical, frictional and corrosion resistant properties, it can be used as replacement of ceramic or cermet type of materials in fabrication and machining. It is also involved in the production of borated steels and borated stainless steels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12006-84-7
Molecular Formula BFe
Molecular Weight (g/mol) 66.66
MDL Number MFCD00016092
InChI Key UJYBTTFGLAOKMN-UHFFFAOYSA-N
Synonym iron boride, iron boride feb, ferrous boride, ironylidyneborane, ferrioborane, ferric boride
PubChem CID 82789
SMILES [B+]=[Fe-]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name ferrioborane
Formula Weight 66.66
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 7.15
Density 7.15
Chemical Name or Material Iron boride
EINECSNumber 234-489-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only