Learn More
Invitrogen™ Select Agar™, powder
Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications.
Supplier: Invitrogen™ 30391023
Description
Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications. Solid culture medium is created by adding agar to media broth (typical final concentration is 1–2%). Solid media cell culture allows the isolation of individual E. coli colonies and is fundamental in cloning workflows.
Agar is used in diagnostic laboratories to detect pathogens from contaminated food or water. Also, it can be used to prepare selective media for saccharomyces cerevisiae and other yeasts used in various molecular microbiology procedures (e.g., YPD Agar). In addition, agar has been used in tissue double embedding for histopathology, hydrogel creation, and tissue engineering applications.
Primary applications
• Making solid media to grow cells (e.g., bacteria, fungi, yeast)
• To isolate E. coli clones with plasmid on a selective media
• Tissue engineering, hydrogel production
Look for more media that fit your needs
We offer other ready-to-use media formulations, such as LB (Lennox) (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Ozone-Safe or Safe for the Ozone Layer
Specifications
• 1 x 500 g bottle Select Agar Powder Store at room temperature. |
|
Select Agar | |
Dehydrated Culture Media | |
500 g | |
Escherichia coli, Saccharomyces cerevisiae |
Bottle | |
Autoclave | |
Coliforms, Fungi | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.