missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Select Agar™, powder

Catalog No. p-4918435
Encompass
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30391023 500 g
30391049 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30391023 Supplier Invitrogen™ Supplier No. 30391023

Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications.

Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications. Solid culture medium is created by adding  agar to media broth (typical final concentration is 1–2%). Solid media cell culture allows the isolation of individual E. coli colonies and is fundamental in cloning workflows.

Agar is used in diagnostic laboratories to detect pathogens from contaminated food or water. Also, it can be used to prepare selective media for saccharomyces cerevisiae and other yeasts used in various molecular microbiology procedures (e.g., YPD Agar). In addition, agar has been used in tissue double embedding for histopathology, hydrogel creation, and tissue engineering applications.

Primary applications
• Making solid media to grow cells (e.g., bacteria, fungi, yeast)
• To isolate E. coli clones with plasmid on a selective media
• Tissue engineering, hydrogel production

Look for more media that fit your needs
We offer other ready-to-use media formulations, such as LB (Lennox) (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage • 1 x 500 g bottle Select Agar Powder

Store at room temperature.

Format Bottle
Media Type Select Agar
Preparation Method Autoclave
Product Type Dehydrated Culture Media
Target Organism Class Coliforms, Fungi
Quantity 500 g
Shipping Condition Room Temperature
Species Escherichia coli, Saccharomyces cerevisiae

For Research Use Only. Not for use in diagnostic procedures.