Learn More
Intertek⢠MIC Test Kit, Petroleum Microbiology Media: TGB
Thioglycollate Medium, for anaerobic and facultative anaerobic heterotrophic bacteria
Supplier: Intertek⢠MK121B
Description
For use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
NACE, TM0194-94 | |
Microbiology Media | |
Vial |
United States | |
Cultivation of heterotrophic anaerobic bacteria, MIC testing | |
288 Vials |