Learn More
Intertek™ MIC Test Kit, Petroleum Microbiology Media: TGB
Catalog No. p-4907874
Encompass
Change view
Click to view available options
Quantity:
12 Vials
288 Vials
Product Type:
Microbiology Media
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
08629006B 12 Vials Microbiology Media
08629006C 12 Vials Microbiology Media
08629006A 12 Vials Microbiology Media
08629013A 288 Vials Microbiology Media
08629013B 288 Vials Microbiology Media
08629013C 288 Vials Microbiology Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 08629006B Supplier Intertek™ Supplier No. MK120B
Only null left
Thioglycollate Medium, for anaerobic and facultative anaerobic heterotrophic bacteria
For use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Certifications/Compliance NACE, TM0194-94
Manufacturing Origin United States
Product Type Microbiology Media
For Use With (Application) Cultivation of heterotrophic anaerobic bacteria, MIC testing
Packaging Type Vial
Quantity 12 Vials