Learn More
Interlocking Cube Grams
Each cube measures one cubic centimeter and weighs one gram.
Supplier:  Customer Channels Group (Ccg) 481800
 View more versions of this product
Catalog No. S94317
Only null left
Explore available promotions
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Description
Description
Ten brightly colored cubes have receptacles and fittings to easily chain together.
Demonstrate the following concepts:
  • Determining conservation of mass
  • Factoring
  • Squaring fractions
  • Pythagorean Theorem
  • Commutative and associative properties
Specifications
Specifications
200/Pk.
Interlocking Centimeter Cube
0.002 lb.
To demonstrate determining conservation of mass, factoring, squaring fractions, Pythagorean theorem, commutative and associative properties
Ten brightly colored cubes with receptacles and fittings
0.9 g
Product Suggestions
Product Suggestions