missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interference Check Standard 5 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$931.74 - $1501.53
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
INTER5-100
|
SPEX CertiPrep Group LLC
INTER5100 |
125 mL | Triple-leached LDPE bottle |
Each for $931.74
|
|
|||||
INTER5-500
|
SPEX CertiPrep Group LLC
INTER5500 |
500 mL | Triple-leached LDPE bottle |
Each for $1,501.53
|
|
|||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ca | 6000ÎĽg/mL |
Fe | 5000ÎĽg/mL |
Mg | 3000ÎĽg/mL |
Al | 1200ÎĽg/mL |
Na | 1000ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
MFCD00011349 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
5% HNO3 | |
63.01 | |
944 | |
Environmental | |
Interference Check Standard 5 |
7697-37-2 | |
HNO3 | |
AA, ICP | |
Certified Reference Material | |
O[N+]([O-])=O | |
nitric acid | |
125 mL | |
CHEBI:48107 | |
Components in 5% HNO3: Al •1200μg/mL Ca •6000μg/mL Fe •5000μg/mL Mg •3000μg/mL Na •1000μg/mL |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only