Learn More
Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter with compatible connector.
Supplier: Mettler Toledo™ 30281896
Description
- Detachable
- Resuable
Specifications
MultiPin-BNC/RCA (cinch) | |
To connect Mettler Toledo InLab sensors to any meter | |
1.2 m |
Mettler Toledo InLab Sensors | |
4 ft. |