Learn More
Invitrogen™ Human TTC24 Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP94527
Description
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56262 (PA5-56262. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
- Environmental benefits include:
- Recyclable
Specifications
A2A3L6 | |
Blocking Assay, Control | |
164118 | |
100 ÎĽL | |
tetratricopeptide repeat domain 24; tetratricopeptide repeat protein 24; TPR repeat protein 24; TTC24 | |
TTC24 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human TTC24 Control Fragment | |
RUO | |
TTC24 | |
Unconjugated | |
Recombinant | |
QVLEKSRRLAERSTGRRLLGHLYNDLGLGYSQLQLFPLAVEAFLQALPLCWVPGEQATVLRNLGMAHNALGNYQEAREFHQKAADLHGSVG | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |