Learn More
Invitrogen™ Human TP53I11 (aa 82-101) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101957
Description
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63770 (PA5-63770. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TP53I11 is a 177 amino acid tumor suppressor belonging to the p53-induced protein gene (PIG) family. The PIG gene family encodes redox-controlling proteins that are involved in p53 tumor suppressor activity. It is suggested that PIG11 is involved in arsenic trioxide As(2)O(3)-induced apoptosis in certain cell lines and may play a significant role in tumor suppression through promotion of cell apoptosis. The gene encoding PIG11 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome.Specifications
| O14683 | |
| Blocking Assay, Control | |
| 9537 | |
| 100 ÎĽL | |
| p53-induced gene 11 protein; Pig11; TP53I11; transformation related protein 53 inducible protein 11; Trp53i11; tumor protein p53 inducible protein 11; tumor protein p53-inducible protein 11 | |
| TP53I11 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human TP53I11 (aa 82-101) Control Fragment | |
| RUO | |
| TP53I11 | |
| Unconjugated | |
| Recombinant | |
| AFPDQLYDAVFDGAQVTSKT | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |