Learn More
Invitrogen™ Human TLL1 (aa 30-57) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP107028
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66344 (PA5-66344. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. A similar protein in mice is required during heart development and specifically processes procollagen C-propeptides and chordin at similar cleavage sites.- Environmental benefits include:
- Renewable Energy
Specifications
O43897 | |
Blocking Assay, Control | |
7092 | |
100 ÎĽL | |
ASD6; TLL; TLL1; tolloid like 1; tolloid-like 1; tolloid-like protein 1 | |
TLL1 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human TLL1 (aa 30-57) Control Fragment | |
RUO | |
TLL1 | |
Unconjugated | |
Recombinant | |
GLDYDYTFDGNEEDKTETIDYKDPCKAA | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |