Learn More
Invitrogen™ Human POTEE (aa 691-714) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP104357
Description
Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60301 (PA5-60301. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein remains unknown.Specifications
Q6S8J3 | |
Blocking Assay, Control | |
445582 | |
100 ÎĽL | |
A26C1; A26C1A; ANKRD26-like family C member 1 A; ANKRD26-like family C, member 1 A; cancer/testis antigen family 104, member 2; CT104.2; POTE ankyrin domain family member E; POTE ankyrin domain family, member E; POTE2; POTE-2; POTE2gamma; POTEE; Prostate, ovary, testis-expressed protein on chromosome 2; protein expressed in prostate, ovary, testis, and placenta 2 | |
POTEE | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human POTEE (aa 691-714) Control Fragment | |
RUO | |
POTEE | |
Unconjugated | |
Recombinant | |
LKALQLNELTMDDDTAVLVIDNGS | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |