Learn More
Invitrogen™ Human OR4C3 Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100604
Description
Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61886 (PA5-61886. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.- Environmental benefits include:
- Renewable Energy
Specifications
Q8NH37 | |
Blocking Assay, Control | |
256144 | |
100 ÎĽL | |
Olfactory receptor 4C3; olfactory receptor family 4 subfamily C member 3; olfactory receptor OR11-98; olfactory receptor, family 4, subfamily C, member 3; OR11-98; OR4C3 | |
OR4C3 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human OR4C3 Control Fragment | |
RUO | |
OR4C3 | |
Unconjugated | |
Recombinant | |
SVTLESMEIPHNITEFVMLGLSQNSEVQR | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |