Learn More
Invitrogen™ Human MYADML2 (aa 1-20) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100257
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61749 (PA5-61749. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MYADML2 is a protein coding gene.Specifications
A6NDP7 | |
Blocking Assay, Control | |
255275 | |
100 ÎĽL | |
1110012N22Rik; Myadml2; myeloid associated differentiation marker like 2; myeloid-associated differentiation marker-like 2; myeloid-associated differentiation marker-like protein 2; RGD1311499 | |
MYADML2 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human MYADML2 (aa 1-20) Control Fragment | |
RUO | |
MYADML2 | |
Unconjugated | |
Recombinant | |
MGSTMEPPGGAYLHLGAVTS | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |