Learn More
Invitrogen™ Human MTX3 Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100242
Description
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61608 (PA5-61608. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Could function in transport of proteins into the mitochondrion.- Environmental benefits include:
- Recyclable
Specifications
Q5HYI7 | |
Blocking Assay, Control | |
345778 | |
100 μL | |
4930470O13Rik; AA409304; AI853833; AU067765; EG624619; Gm1194; Gm6514; metaxin 3; metaxin-3; mtx3; pleckstrin homology domain containing, family M (with RUN domain) member 2; zgc:109718; zMTX3 | |
MTX3 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human MTX3 Control Fragment | |
RUO | |
MTX3 | |
Unconjugated | |
Recombinant | |
SESLVVMAYAKFSGAPLKVNVIDNTWRGSRGDVPILTTEDDMVSQPG | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |