Learn More
Invitrogen™ Human HMGB4 (aa 96-173) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP94470
Description
Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57370 (PA5-57370. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.Specifications
Q8WW32 | |
Blocking Assay, Control | |
127540 | |
100 ÎĽL | |
1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein | |
HMGB4 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human HMGB4 (aa 96-173) Control Fragment | |
RUO | |
HMGB4 | |
Unconjugated | |
Recombinant | |
PPSSFLLFCQDHYAQLKRENPNWSVVQVAKATGKMWSTATDLEKHPYEQRVALLRAKYFEELELYRKQCNARKKYRMS | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |