Learn More
Invitrogen™ Human ELF4 (aa 331-409) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101163
Description
Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63593 (PA5-63593. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5'-WGGA-3'. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.- Environmental benefits include:
- Recyclable
- Recycled Content
- Renewable Materials
Specifications
Q99607 | |
Blocking Assay, Control | |
2000 | |
100 ÎĽL | |
AV314029; BC042423; E74 like ETS transcription factor 4; E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ELFR; ENSMUSG00000053512; ETS-related transcription factor Elf-4; Gm9907; MEF; myeloid elf-1 like factor; myeloid Elf-1-like factor; RGD1560743 | |
Elf4 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human ELF4 (aa 331-409) Control Fragment | |
RUO | |
ELF4 | |
Unconjugated | |
Recombinant | |
SSRVSSRSAPQGKGSSSWEKPKIQHVGLQPSASLELGPSLDEEIPTTSTMLVSPAEGQVKLTKAVSASSVPSNIHLGVA | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |