Learn More
Invitrogen™ Human C9orf64 (aa 255-340) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP105386
Description
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66828 (PA5-66828. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Involved in salvaging queuosine.Specifications
Q5T6V5 | |
Blocking Assay, Control | |
84267 | |
100 ÎĽL | |
C9orf64; chromosome 9 open reading frame 64; Queuosine salvage protein; UPF0553 protein; UPF0553 protein C9orf64 | |
C9orf64 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human C9orf64 (aa 255-340) Control Fragment | |
RUO | |
C9orf64 | |
Unconjugated | |
Recombinant | |
SDDLLKKLLKGEMLSYGDRQEVEIRGCSLWCVELIRDCLLELIEQKGEKPNGEINSILLDYYLWDYAHDHREDMKGIPFHRIRCIY | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |