missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Howard Leight™ AirSoft™ Ear Plugs

AirSoft™ four-flange design creates a secure seal in the ear canal, delivering optimal protection and increased comfort for long-term wear.

Supplier:  Honeywell™ AS1

 View more versions of this product

Catalog No. 19-013972



Only null left
Explore available promotions

Description

Description

  • Advanced air pocket design features internal noise-blocking fins
  • Four-flange profile creates secure seal and gives a sense of less pressure in the ear canal
  • Rounded flanges, tapered shape fits securely
  • Firm stem facilitates easy insertion and removal
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications

Specifications

Yes
N/A
Universal
No
100 Pairs/Pk.
Blue
Uncorded
Reusable
Uncorded
Class AL
Construction, Energy and Power, Manufacturing, Mining, Oil and Gas, Transportation, and Other Various Industries
Poly bag
TPE
27 dB
Flanged
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions