missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Howard Leight™ AirSoft™ Ear Plugs

AirSoft™ four-flange design creates a secure seal in the ear canal, delivering optimal protection and increased comfort for long-term wear.

$72.69 - $150.26

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy and Power, Manufacturing, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility No
View More Specs

Products 4
Catalog Number Mfr. No. Corded Cord Material Packaging Type Price Quantity  
Catalog Number Mfr. No. Corded Cord Material Packaging Type Price Quantity  
19-013978
Accessories View Documents Promotion Details
Honeywell™
DPAS30R
Corded Vinyl Flip-top box
Pack of 100 Pair for $150.26
Case of 10 Pack for $1,364.20
 
18-534
Accessories View Documents Promotion Details
Honeywell™
DPAS30W
Corded Nylon Flip-top box
Pack of 100 Pair for $140.18
Case of 10 Pack for $1,272.70
 
19-013-976
Accessories View Documents Promotion Details
Honeywell™
AS30W
Corded Nylon Safety impact case
Pack of 50 Pair for $97.02
Case of 10 Pack for $880.90
 
19-013972
View Documents Promotion Details
Honeywell™
AS1
Uncorded N/A Poly bag
Pack of 50 Pair for $72.69
Case of 10 Pack for $660.00
 
Description

Description

  • Advanced air pocket design features internal noise-blocking fins
  • Four-flange profile creates secure seal and gives a sense of less pressure in the ear canal
  • Rounded flanges, tapered shape fits securely
  • Firm stem facilitates easy insertion and removal
Specifications

Specifications

Yes
Universal
No
Blue
Flanged
No
Class AL
Construction, Energy and Power, Manufacturing, Mining, Oil and Gas, Transportation, and Other Various Industries
TPE
27 dB
Reusable
SDS