Learn More
Hoeferâ„¢ SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarine Combs
Accessories for SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarines
$179.33 - $185.43
Specifications
For Use With (Equipment) | SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine |
---|---|
Type | Comb |
Depth (English) Pan | 0.19 in. |
Depth (Metric) Pan | 0.5 cm |
Catalog Number | Mfr. No. | No. of Wells | Thickness (Metric) | Width (Metric) Well | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Wells | Thickness (Metric) | Width (Metric) Well | Price | Quantity | ||||
03-500-780
![]() |
Hoeferâ„¢
SUB20C1.520MC |
20 | 0.15 cm | 0.7 cm |
Each for $182.37
|
|
||||
03-500-786
![]() |
Hoeferâ„¢
SUB20C1.540MC |
40 | 0.15 cm | 0.3 cm |
Each for $185.43
|
|
||||
03-500-776
![]() |
Hoeferâ„¢
SUB20C116 |
16 | 0.1 cm | 0.85 cm |
Each for $185.43
|
|
||||
03-500-779
![]() |
Hoeferâ„¢
SUB20C120MC |
20 | 0.1 cm | 0.7 cm |
Each for $185.43
|
|
||||
03-500-782
![]() |
Hoeferâ„¢
SUB20C128 |
28 | 0.1 cm | 0.5 cm |
Each for $185.43
|
|
||||
03-500-785
![]() |
Hoeferâ„¢
SUB20C140MC |
40 | 0.1 cm | 0.3 cm |
Each for $179.33
|
|
||||
03-500-778
![]() |
Hoeferâ„¢
SUB20C216 |
20 | 0.2 cm | 0.85 cm |
Each for $182.37
|
|
||||
03-500-781
![]() |
Hoeferâ„¢
SUB20C220MC |
20 | 0.2 cm | 0.7 cm |
Each for $179.33
|
|
||||
03-500-784
![]() |
Hoeferâ„¢
SUB20C228 |
28 | 0.2 cm | 0.5 cm |
Each for $179.33
|
|
||||
03-500-787
![]() |
Hoeferâ„¢
SUB20C240MC |
40 | 0.2 cm | 0.3 cm |
Each for $179.33
|
|
||||
Description
- Ranges from 16, 20, 28 and 40 Wells
- 1, 1.5 and 2 mm Thickness
- 3, 5, 7 and 8.5 mm Well Width
- Min. 13, max. 75 μL Sample Volume
Specifications
SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine | |
Comb | |
0.19 in. | |
0.5 cm |