Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Manufacturer:  Hoefer™ HE91A201.5
 View more versions of this product
Catalog No. 36-100-3930
Requires : Comb back must be ordered separately (see ordering table)
Description
Description
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Combs can be used interchangeably with a single comb back.
Specifications
Specifications
0.19 in.
HE99X Max Submarine Electrophoresis Unit
20
0.15 cm
0.18 in.
0.5 cm
Backless Comb
0.05 in.
35.5 μL
0.47 cm