Learn More
Hematoxylin Stain Solution, Gill 2 Formulation, Double Strength, Ricca Chemical
For More Intense Nuclei in Progressive Cytology and Delicate Nuclei in Progressive Histology
Supplier:  Ricca Chemical Company 353616
 View more versions of this product
Catalog No. 3536-16
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Hematoxylin Stain Solution
7732-18-5,107-21-1,64-19-7,7784-31-8,517-28-2,7681-55-2
75.0,20.88,3.15,2.64,0.3,0.03
Natural Poly Bottle
2 to 4
Liquid
Gill 2 Formulation, Double Strength
72.0,20.06,3.03,2.54,0.29,0.03
Laboratory
Dark Purple
500 mL
Product Suggestions
Product Suggestions