missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hematoxylin Stain Solution, Gill 2 Formulation, Double Strength, Ricca Chemical
For More Intense Nuclei in Progressive Cytology and Delicate Nuclei in Progressive Histology
$202.48 - $545.13
Specifications
Chemical Name or Material | Hematoxylin Stain Solution |
---|---|
Name Note | Gill 2 Formulation, Double Strength |
CAS Min % | 72.0,20.06,3.03,2.54,0.29,0.03 |
CAS Max % | 75.0,20.88,3.15,2.64,0.3,0.03 |
CAS | 107-21-1,64-19-7,7784-31-8,517-28-2,7681-55-2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
3536-16
|
Ricca Chemical Company
353616 |
500 mL | Natural Poly Bottle |
Each for $202.48
|
|
||||
3536-32
|
Ricca Chemical Company
353632 |
1 L | Natural Poly Bottle |
Each for $363.84
|
|
||||
35-365
|
Les Produits Chimiques
8255725 |
20 L | Cubitainer |
Each for $545.13
|
|
||||
Specifications
Hematoxylin Stain Solution | |
72.0,20.06,3.03,2.54,0.29,0.03 | |
107-21-1,64-19-7,7784-31-8,517-28-2,7681-55-2 | |
Liquid |
Gill 2 Formulation, Double Strength | |
75.0,20.88,3.15,2.64,0.3,0.03 | |
Purple |