missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaraldehyde, 25% aqueous solution, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219519910
Specifications
Glutaraldehyde | |
Colorless | |
C5H8O2 | |
10 mL | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.11 |
111-30-8 | |
1.061g/mL | |
MFCD00007025 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
Liquid |
Chemical Identifiers
111-30-8 | |
100.12 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
3485 | |
pentanedial |
C5H8O2 | |
MFCD00007025 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehydeShow More | |
CHEBI:64276 | |
O=CCCCC=O |