missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, 25% aqueous solution, MP Biomedicals™

Catalog No. p-4405801
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19519910 10 mL
ICN19519980 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19519910 Supplier MP Biomedicals Inc Supplier No. 0219519910
Only null left

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Color Colorless
Density 1.061g/mL
Quantity 10 mL
Formula Weight 100.11
Physical Form Liquid
Chemical Name or Material Glutaraldehyde