missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Soln., 4%, Buffered, pH 6.9 (approx. 10% Formalin soln.), For Histology, MilliporeSigma™
Supplier: Technidata 1004965000
Description
For histologySpecifications
Liquid | |
approx. 10% Formalin solution | |
Characteristic odor | |
Colorless | |
4% |
Formaldehyde solution | |
50-00-0,67-56-1 | |
Plastic container | |
5 L | |
Formaline solution, Methanal solution |