missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Soln., 4%, Buffered, pH 6.9 (approx. 10% Formalin soln.), For Histology, MilliporeSigma™
$261.63 - $477.89
Specifications
| Chemical Name or Material | Formaldehyde solution |
|---|---|
| Physical Form | Liquid |
| Name Note | approx. 10% Formalin solution |
| CAS | 50-00-0,67-56-1 |
| Odor | Characteristic odor |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M1004965000
|
Technidata
1004965000 |
5 L |
Each for $261.63
|
|
|||||
|
M1004969010
|
Technidata
1004969010 |
10 L |
Each for $477.89
|
|
|||||
Description
For histologySpecifications
| Formaldehyde solution | |
| approx. 10% Formalin solution | |
| Characteristic odor | |
| Colorless | |
| Formaline solution, Methanal solution |
| Liquid | |
| 50-00-0,67-56-1 | |
| Plastic container | |
| 4% |