Learn More
Foamtec UltraMOP™ Self-Wringing Mop Heads
Clean room sponge mop head refill for Roll-O-Matic™
Supplier: Foamtec FS600
Description
- Tear-resistant, highly absorbent polyurethane sponge
- With stainless steel bracket
- Pin-hole-free foam for abrasion resistance
- Available in Bump-Proof™ micro fiber over foam versions
- For wet mopping procedures in sterile and controlled environments
Specifications
14 in. | |
Autoclavable | |
Designed for wet mopping procedures in sterile and controlled environments |
Non-sterile | |
Stainless steel bracket |