missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accuSkan™ FC Filter-Based Microplate Photometer
For research and routine ELISA applications
$15311.74 - $21189.43
Specifications
Detector Type | Photodetector |
---|---|
Dimensions (L x W x H) | 11.4 x 15.7 x 8.7 in. (29 x 40 x 22 cm) |
Interface | PC Control, Internal Software |
Light Source | Quartz Halogen Lamp |
Measurement Speed | 96 wells: 6 sec. (fast mode), 12 sec. (normal mode) |
Catalog Number | Mfr. No. | Description | Plate Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Plate Type | Price | Quantity | ||||
14-377-575
|
Fisherbrand™
51119040 |
accuSkan FC | 96-well Plate |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14-377-576
|
Fisherbrand™
51119140 |
accuSkan FC with incubator | 96 and 384-well Plates |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
A proven ELISA solution—accuSkan FC and the Fisher Scientific accuWash washer. The combination of the accuSkan FC microplate photometer and the accu-Wash™ microplate washer offers a convenient modular solution for routine and research ELISA applications.
- Broad wavelength range of 340 to 850nm for a wide variety of applications
- Eight-position filter wheel with three standard pre-installed filters (405, 450 and 620nm)
- Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
- On-board linear shaking and optional incubation up to 50°C for temperature critical assays
- Superior usability with both internal software and dedicated PC software
- Data export capability via USB memory stick for easy data transfer
Warranty
1 Year
Specifications
Photodetector | |
PC Control, Internal Software | |
96 wells: 6 sec. (fast mode), 12 sec. (normal mode) | |
±1% (0 to 3 Abs) or ±0.003 Abs, which ever is greater at 405 nm | |
8-position filter wheel; filters installed: 405nm, 450nm, and 620nm | |
96-well plate: 0 to 3 Abs (±2% at 405 nm) 384-well plate: 0 to 2.5 Abs (±2% at 405 nm) |
|
0.001 Abs | |
340 to 850 nm |
11.4 x 15.7 x 8.7 in. (29 x 40 x 22 cm) | |
Quartz Halogen Lamp | |
Linear shaking with 3 modes | |
100/240 V, 50/60 Hz | |
Robotic automation | |
CV ≤ 0.2% (0.3 to 3 Abs) at 405 nm | |
1 Year |