missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Healthcare™ PROTOCOL™ Color-Coded Prefilled Formalin Containers
Prefilled with 10% neutral buffered formalin
$106.42 - $129.10
Specifications
Graduated | Yes |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Closure Type | Color-coded Cap with Leakproof O-Ring |
Label | Paper |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Price | Quantity | ||||
23-316156
|
Fisher Healthcare™
316156 |
90 mL, 45 mL Formalin fill | Orange |
Case of 75 Each for $129.10
|
|
||||
23-032060
|
Fisher Healthcare™
032060 |
120 mL, 60 mL Formalin fill | Yellow |
Case of 50 Each for $106.42
|
|
||||
23-316155
![]() |
Fisher Healthcare™
316155 |
60 mL, 30 mL Formalin fill | Blue |
Case of 75 Each for $118.00
|
![]() |
||||
Description
- Color-coded screw cap with leakproof O-ring
- Autoclavable polypropylene
- Paper labels to indicate date, doctor, patient name or ID, and specimen type
- Graduated
Order Info
The 20/10, 40/20 and 60/30mL containers are packaged in shrink wrapped trays to allow for ease of use, handling and storage.
Specifications
Yes | |
Autoclavable | |
Paper |
Polypropylene | |
Color-coded Cap with Leakproof O-Ring |