missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ PROTOCOL™ Color-Coded Prefilled Formalin Containers

Catalog No. 23316156
Change view
Click to view available options
Capacity (Metric):
90 mL, 45 mL Formalin fill
Closure Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
23316156 90 mL, 45 mL Formalin fill Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23316156 Supplier Fisher Healthcare™ Supplier No. 316156
Only null left

Prefilled with 10% neutral buffered formalin

  • Color-coded screw cap with leakproof O-ring
  • Autoclavable polypropylene
  • Paper labels to indicate date, doctor, patient name or ID, and specimen type
  • Graduated

Order Info

The 20/10, 40/20 and 60/30mL containers are packaged in shrink wrapped trays to allow for ease of use, handling and storage.

Specifications

Graduated Yes
Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 90 mL, 45 mL Formalin fill
Closure Color Orange
Closure Type Color-coded Cap with Leakproof O-Ring
Label Paper