Learn More
Thermo Scientific™ GeneRuler High Range DNA Ladder
Perform fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels.
Supplier: Thermo Scientific™ SM1352
Description
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5hour, 0.4%agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol in the Resources.)
The ladder is supplied with 6X DNA Loading Dye for sample DNA.
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5 hours, 0.4% agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.The ladder is supplied with 6X DNA Loading Dye for sample DNA.- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Supplied with loading dye for sample DNA
- Ideal for fast sizing of high molecular weight DNA fragments
- Environmental benefits include:
- Recyclable
Specifications
6X | |
Xylene Cyanol FF, Orange G | |
No | |
Agarose Gels | |
GeneRuler |
Agarose | |
High Range DNA Ladder | |
10171 to 48502 bp | |
Sustainable packaging | |
5 x 50 μg |
For Research Use Only. Not for use in diagnostic procedures.