missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler High Range DNA Ladder
GREENER_CHOICE

Catalog No. p-4529759
Change view
Click to view available options
Quantity:
5 x 50 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSM1351 50 μg
FERSM1352 5 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERSM1351 Supplier Thermo Scientific™ Supplier No. SM1351
Only null left

Perform fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels.

GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5hour, 0.4%agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol in the Resources.)

The ladder is supplied with 6X DNA Loading Dye for sample DNA.

GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5 hours, 0.4% agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.The ladder is supplied with 6X DNA Loading Dye for sample DNA.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Supplied with loading dye for sample DNA
  • Ideal for fast sizing of high molecular weight DNA fragments

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Size Range 10171 to 48502 bp
Gel Compatibility Agarose Gels
Quantity 50 μg
Product Line GeneRuler
Product Type High Range DNA Ladder
Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.

For Research Use Only. Not for use in diagnostic procedures.