Learn More
ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 4815-37-6 |
---|---|
Molecular Formula | C14H15NO2S |
Molecular Weight (g/mol) | 261.339 |
MDL Number | MFCD01763475 |
InChI Key | SXONDAWSQJWZEO-UHFFFAOYSA-N |
Synonym | 2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl esterShow More |
PubChem CID | 693902 |
IUPAC Name | ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate |
SMILES | CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AW00937DA
|
Thermo Scientific Chemicals
AW00937DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AW00937DE
|
Thermo Scientific Chemicals
AW00937DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AW00937EB
|
Thermo Scientific Chemicals
AW00937EB |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
4815-37-6 | |
261.339 | |
SXONDAWSQJWZEO-UHFFFAOYSA-N | |
693902 | |
CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N |
C14H15NO2S | |
MFCD01763475 | |
2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl esterShow More | |
ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate |
Specifications
4815-37-6 | |
100.0 | |
Amber glass bottle | |
MFCD01763475 | |
2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester | |
CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N | |
261.339 | |
261.34 | |
ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate |
96.5 | |
97% | |
C14H15NO2S | |
1g | |
SXONDAWSQJWZEO-UHFFFAOYSA-N | |
ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate | |
693902 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning