missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW00937DA

 View more versions of this product

Catalog No. AW00937DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

ethyle2-amino-5-methyl-4-phenylthiophene-3-carboxylate
96.5
97%
C14H15NO2S
1g
SXONDAWSQJWZEO-UHFFFAOYSA-N
ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
693902
97%
4815-37-6
100.0
Amber glass bottle
MFCD01763475
2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester
CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N
261.339
261.34

Chemical Identifiers

4815-37-6
261.339
SXONDAWSQJWZEO-UHFFFAOYSA-N
693902
CCOC(=O)C1=C(SC(=C1C2=CC=CC=C2)C)N
C14H15NO2S
MFCD01763475
2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, ethyl ester, maybridge1_000503, upcmld-dp080, upcmld-dp080:001, ethyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 3-thiophenecarboxylic acid,2-amino-5-methyl-4-phen, 2-amino-4-phenyl-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-4-phenyl-, ethyl ester
ethyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions