missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hexafluoroglutaryl chloride, 97%, Thermo Scientific Chemicals

$640.81 - $640.81

Chemical Identifiers

CAS 18381-53-8
Molecular Formula C7H5ClF6O3
Molecular Weight (g/mol) 286.55
MDL Number MFCD00054671
InChI Key OLRXGDHRDQKNGW-UHFFFAOYSA-N
PubChem CID 161251
IUPAC Name ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1404906
View Documents
Thermo Scientific Chemicals
L1404906
5 g
Each for $640.81
Only null left
 
Description

Description

Ethyl hexafluoroglutaryl chloride is used as a derivatising agent in gas chromatography. It is an active reagent used for the quantization of amphetamine and methamphetamine in urine by gas chromatography/mass spectroscopy (GC/MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hexafluoroglutaryl chloride is used as a derivatising agent in gas chromatography. It is an active reagent used for the quantization of amphetamine and methamphetamine in urine by gas chromatography/mass spectroscopy (GC/MS).

Solubility
Miscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases and water.
Specifications

Chemical Identifiers

18381-53-8
286.55
OLRXGDHRDQKNGW-UHFFFAOYSA-N
ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
C7H5ClF6O3
MFCD00054671
161251
CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O

Specifications

18381-53-8
158°C to 160°C
1.359
5 g
1884844
4-carbethoxyhexafluorobutyryl chloride, ethyl hexafluoroglutaryl chloride, pentanoic acid, 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxo-, ethyl ester, 4ce-hfba, 4-carbethoxyhexafluorobutyryl, ethyl hexafluoroglutaryl chloride 250mg, 4-ethoxycarbonyl-4,4,3,3,2,2-hexafluorobutyryl chloride, pentanoic acid,5-chloro-2,2,3,3,4,4-hexafluoro-5-oxo-, ethyl ester
OLRXGDHRDQKNGW-UHFFFAOYSA-N
ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
161251
97%
1.47
C7H5ClF6O3
MFCD00054671
UN3265
Moisture sensitive
Miscible with water.
CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O
286.55
286.56
Ethyl hexafluoroglutaryl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only