Learn More
Ethyl 4-bromobenzoate, 99%
CAS: 5798-75-4 | C9H9BrO2 | 229.07 g/mol
Supplier: Thermo Scientific Chemicals 249760500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 4-bromobenzoate | |
5798-75-4 | |
1.4300g/mL | |
>110°C | |
Glass bottle | |
1.5440 to 1.547 | |
MFCD00016329 | |
1.43 | |
Solubility in water: slightly soluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents | |
CCOC(=O)C1=CC=C(Br)C=C1 | |
229.07 | |
229.06 |
99% | |
18°C | |
262°C to 264°C | |
98.5% min. (GC) | |
C9H9BrO2 | |
BrC6H4CO2CH2CH3 | |
50 mL | |
4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoate | |
XZIAFENWXIQIKR-UHFFFAOYSA-N | |
ethyl 4-bromobenzoate | |
22043 | |
99% |
Chemical Identifiers
5798-75-4 | |
229.07 | |
XZIAFENWXIQIKR-UHFFFAOYSA-N | |
22043 | |
CCOC(=O)C1=CC=C(Br)C=C1 |
C9H9BrO2 | |
MFCD00016329 | |
4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoateShow More | |
ethyl 4-bromobenzoate |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 227-347-2
TSCA : TSCA
RUO – Research Use Only