missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERBB2 Rabbit anti-Human, Unconjugated, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation DH0082
Description
ERBB2 Monoclonal antibody specifically detects ERBB2 in Human samples. It is validated for IF.Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
ERBB2 | |
Monoclonal | |
Unconjugated | |
ERBB2 | |
Rabbit | |
12 ÎĽL | |
2064 | |
Store at -20°C. Do not freeze. | |
IgG |
Immunofluorescence | |
0.2 mg/mL | |
v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
CD340, HER-2, HER-2/neu, HER2, NEU, NGL, TKR1 | |
Human recombinant ERBB2 | |
Primary | |
Human | |
Liquid |