Learn More
Agilent BioTek Empty Filter Wheel for Synergy and FLx800 Microplate Reader
GREENER_CHOICE
For use with FLx800, Synergy 2, Synergy H4 and Synergy HT microplate readers.
Supplier:  Agilent BioTek 7080541
Catalog No. BT7080541
Only null left
Explore available promotions
Description
Description
  • Can hold up to 4 filters
  • EX and EM labels provided
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
EX and EM labels
Product Suggestions
Product Suggestions