Learn More
MilliporeSigma™ Vent Filters S.D.S Systems (purchased after 12/30/03)
Protect high-resistivity water product from ingress of possible airborne contaminants from the environment into S.D.S tank during storage
Supplier:  MilliporeSigma™ TANKVNT01
 View more versions of this product
Catalog No. TANKVNT01
Only null left
Explore available promotions
Description
Description
Protect high-resistivity water product from ingress of possible airborne contaminants from the environment into S.D.S tank during storage
  • Filter incorporates activated carbon to absorb volatile organics, soda lime bed to remove carbon dioxide, and an absolute membrane to remove particles and bacteria from the laboratory atmosphere
  • Filters easily install in built-in holder on front of S.D.S. reservoir
Note:These filters are for use with S.D.S. systems purchased after December 31, 2003; for S.D.S systems purchased before December 31, 2003, contact your Fisher Scientific™ Customer Service Representative
Specifications
Specifications
Vent Filter
RiOs™ system
Vent Filter
Product Suggestions
Product Suggestions