Learn More
MilliporeSigma™ Vent Filters S.D.S Systems (purchased after 12/30/03)
Description
Protect high-resistivity water product from ingress of possible airborne contaminants from the environment into S.D.S tank during storage
- Filter incorporates activated carbon to absorb volatile organics, soda lime bed to remove carbon dioxide, and an absolute membrane to remove particles and bacteria from the laboratory atmosphere
- Filters easily install in built-in holder on front of S.D.S. reservoir
Note:These filters are for use with S.D.S. systems purchased after December 31, 2003; for S.D.S systems purchased before December 31, 2003, contact your Fisher Scientific™ Customer Service Representative
Specifications
Specifications
| Type | Vent Filter |
| For Use With (Equipment) | Elix™ system |
| Includes | CO2 trap |
| Model | Vent Filter |