Learn More
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation
Supplier:  MilliporeSigma™ MSFBN6B50
Catalog No. MSFBN6B50
Only null left
Explore available promotions
Description
Description
  • High Throughput
  • Meet ANSI/SBS 2004 plate specifications
  • Protects underdrain, eliminates surface contact with individual well ‘drip directors’
  • Plastic skirt enables stacking directly with collection plates
Applications
  • Receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation
  • Radioisotope detection
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
    • Certifications and Seals of Approval
Specifications
Specifications
ANSI, SBS
Hydrophilic PVDF
1/0.65 μm
Barex
Non-sterile
Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification
0.5 x 8.55 x 0.56 in. (1.28 x 21.71 x 1.44 cm)
21.71 cm
Standard
8 x 12
Opaque
96
0.26 cm2
50/Pk.
1.28 cm
1.44 cm
Product Suggestions
Product Suggestions