Learn More
MilliporeSigmaâ„¢ Millistak+â„¢ Pod Disposable Depth Filter System
Ideal for primary and secondary clarification in lab, pilot and process-scale applications
$334.00 - $502.92
Specifications
Disposable | Single-use |
---|---|
Material | Glass Fiber-reinforced Polypropylene |
For Use With (Application) | Clarification of serum, plasma, vaccines, cell culture or other fluids |
Inlet Connection (English) | 0.5 in. hose barb |
Max. Pressure | 50 psi at 25°C, 15 psi at 80°C |
Catalog Number | Mfr. No. | Grade | Filtration Area | Material (Membrane) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Grade | Filtration Area | Material (Membrane) | Price | Quantity | ||||
MCE15054H1
|
MilliporeSigmaâ„¢
MCE15054H1 |
CE15 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE20027H1
|
MilliporeSigmaâ„¢
MCE20027H1 |
CE20 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE15027H1
|
MilliporeSigmaâ„¢
MCE15027H1 |
CE15 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE20054H1
|
MilliporeSigmaâ„¢
MCE20054H1 |
CE20 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE25027H1
|
MilliporeSigmaâ„¢
MCE25027H1 |
CE25 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE25054H1
|
MilliporeSigmaâ„¢
MCE25054H1 |
CE25 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE30054H1
|
MilliporeSigmaâ„¢
MCE30054H1 |
CE30 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE35054H1
|
MilliporeSigmaâ„¢
MCE35054H1 |
CE35 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE30027H1
|
MilliporeSigmaâ„¢
MCE30027H1 |
CE30 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE40027H1
|
MilliporeSigmaâ„¢
MCE40027H1 |
CE40 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE35027H1
|
MilliporeSigmaâ„¢
MCE35027H1 |
CE35 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE40054H1
|
MilliporeSigmaâ„¢
MCE40054H1 |
CE40 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE50027H1
|
MilliporeSigmaâ„¢
MCE50027H1 |
CE50 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MDE25027H1
|
MilliporeSigmaâ„¢
MDE25027H1 |
DE25 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE25054H1
|
MilliporeSigmaâ„¢
MDE25054H1 |
DE25 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE30054H1
|
MilliporeSigmaâ„¢
MDE30054H1 |
DE30 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE35054H1
|
MilliporeSigmaâ„¢
MDE35054H1 |
DE35 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE35027H1
|
MilliporeSigmaâ„¢
MDE35027H1 |
DE35 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE30027H1
|
MilliporeSigmaâ„¢
MDE30027H1 |
DE30 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE40054H1
|
MilliporeSigmaâ„¢
MDE40054H1 |
DE40 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $502.92
|
|
||||
MDE40027H1
|
MilliporeSigmaâ„¢
MDE40027H1 |
DE40 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE45027H1
|
MilliporeSigmaâ„¢
MDE45027H1 |
DE45 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE50027H1
|
MilliporeSigmaâ„¢
MDE50027H1 |
DE50 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE45054H1
|
MilliporeSigmaâ„¢
MDE45054H1 |
DE45 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE50054H1
|
MilliporeSigmaâ„¢
MDE50054H1 |
DE50 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE60027H1
|
MilliporeSigmaâ„¢
MDE60027H1 |
DE60 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE55054H1
|
MilliporeSigmaâ„¢
MDE55054H1 |
DE55 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE55027H1
|
MilliporeSigmaâ„¢
MDE55027H1 |
DE55 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE65027H1
|
MilliporeSigmaâ„¢
MDE65027H1 |
DE65 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE60054H1
|
MilliporeSigmaâ„¢
MDE60054H1 |
DE60 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE75054H1
|
MilliporeSigmaâ„¢
MDE75054H1 |
DE75 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE70054H1
|
MilliporeSigmaâ„¢
MDE70054H1 |
DE70 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $407.00
|
|
||||
MDE65054H1
|
MilliporeSigmaâ„¢
MDE65054H1 |
DE65 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
Each for $424.00
|
|
||||
MDE70027H1
|
MilliporeSigmaâ„¢
MDE70027H1 |
DE70 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MDE75027H1
|
MilliporeSigmaâ„¢
MDE75027H1 |
DE75 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
Each for $334.00
|
|
||||
MCE45027H1
|
MilliporeSigmaâ„¢
MCE45027H1 |
CE45 | 0.027 m2 | Cellulose |
Each for $334.00
|
|
||||
MCE50054H1
|
MilliporeSigmaâ„¢
MCE50054H1 |
CE50 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
MCE45054H1
|
MilliporeSigmaâ„¢
MCE45054H1 |
CE45 | 0.054 m2 | Cellulose |
Each for $424.00
|
|
||||
Description
- Filter Material: cellulose
- Configurations: 0.11, 0.77, or 1.4 m2 filtration area
- Low hold-up volume for greater product yield
- Broad range of 23 media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000L or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Disposable Pod system protects operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Single-use | |
Clarification of serum, plasma, vaccines, cell culture or other fluids | |
50 psi at 25°C, 15 psi at 80°C | |
Millistak+ Pod Depth Filter | |
5.3 in. |
Glass Fiber-reinforced Polypropylene | |
0.5 in. hose barb | |
1 Ea. | |
5 to 12,000 L | |
13.46 cm |