missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eaton's reagent
CAS: 39394-84-8 | O10P4
$379.89 - $379.89
Specifications
Chemical Name or Material | Eaton's reagent |
---|---|
Color | Undesignated |
CAS | 39394-84-8,75-75-2,1314-56-3 |
Boiling Point | 122.0°C (1.0 mmHg) |
Molecular Formula | O10P4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC369805000
|
Thermo Scientific Chemicals
369805000 |
500 mL |
Each for $379.89
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Eaton's reagent | |
39394-84-8,75-75-2,1314-56-3 | |
O10P4 | |
6.7 to 8.2 % | |
1.5000g/mL | |
283.89 | |
Glass bottle | |
1.5 | |
Liquid |
Undesignated | |
122.0°C (1.0 mmHg) | |
MFCD00070545 | |
05,535; 07,291; 08,400; 11,428 | |
Phosphorus pentoxide, 7.5 wt.% solution in methanesulfonic acid | |
6.7 to 8.2% | |
1.4330 to 1.4360 | |
>110°C |
RUO – Research Use Only