missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Tag Monoclonal Antibody (FG4R), HRP, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen MA191878HRP
Description
similar epitope as the FLAG tag from Sigma MA1-91878 detects Flag tag from various samples. MA1-91878 has been successfully used in Western blot applications. The MA1-91878 immunogen is synthetic peptide (DYKDDDDK) coupled to KLH.
DYKDDDDK tag, also known as FLAG tag, is a short peptide sequence that is commonly used as a protein tag to facilitate protein purification, detection, and localization. The tag consists of eight amino acids: Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys. It is often fused to the N- or C-terminus of a protein of interest, allowing for easy identification and isolation of the protein using anti-FLAG antibodies. The DYKDDDDK tag is widely used in molecular biology and biochemistry research, and has been employed in various applications, such as protein-protein interaction studies, protein expression analysis, and protein localization studies. The FLAG tag structure has been optimized for compatibility with the proteins it is attached to, in that it is more hydrophilic than other common epitope tags and therefore less likely to denature or inactivate proteins to which it is appended. FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany.Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
DYKDDDDK Tag | |
Monoclonal | |
1 mg/mL | |
antibody with 0.07% Kathon | |
Mouse | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG2b |
Western Blot | |
FG4R | |
HRP | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
Synthetic peptide (DYKDDDDK) coupled to KLH. | |
50 ÎĽL | |
Primary | |
4° C | |
Liquid | |
Tag |