Learn More
Doxycycline monohydrate
Doxycycline monohydrate, CAS # 17086-28-1, is the hydrated form of the tetracycline class antibiotic doxycycline. | CAS: 17086-28-1 | C22H26N2O9 | 462.455 g/mol
$79.79 - $300.10
Chemical Identifiers
CAS | 17086-28-1 |
---|---|
Molecular Formula | C22H26N2O9 |
Molecular Weight (g/mol) | 462.455 |
MDL Number | MFCD02682958 |
InChI Key | FZKWRPSUNUOXKJ-CVHRZJFOSA-N |
PubChem CID | 54684461 |
IUPAC Name | (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrateShow More |
SMILES | CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ6380506
|
Thermo Scientific Chemicals
J6380506 |
5 g |
Each for $79.79
|
|
||||||
AAJ6380514
|
Thermo Scientific Chemicals
J6380514 |
25 g |
Each for $300.10
|
|
||||||
Description
Doxycycline monohydrate acts as an antibiotic, antimicrobial agent and a matrix metalloproteinase inhibitor. In addition to this, it inhibits the bacterial protein synthesis by preventing the interaction of aminoacyl-tRNA and the bacterial ribosome.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Doxycycline monohydrate is a semi-synthetic tetracycline antibiotic with broad-spectrum activity
- It is a tetracycline with the 5beta-hydrogen group replaced by a hydroxy group
- Doxycycline monohydrate acts by binding the 30S ribosomal subunit and preventing polypeptide formation
- Doxycycline monohydrate may also act on the 50S ribosomal subunit, is a known inhibitor of collagenases, and has anti-inflammatory properties
Application
- Doxycycline can be used for study of cancers, infections, and antibiotic resistance in laboratory settings
- Doxycycline can be used experimentally to control TET genes and induce expression of genes of interest in vitro
Chemical Identifiers
17086-28-1 | |
462.455 | |
FZKWRPSUNUOXKJ-CVHRZJFOSA-N | |
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrateShow More |
C22H26N2O9 | |
MFCD02682958 | |
54684461 | |
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O |
Specifications
Doxycycline Monohydrate | |
17086-28-1 | |
Yellow | |
C22H26N2O9 | |
14,3440 | |
FZKWRPSUNUOXKJ-CVHRZJFOSA-N | |
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate | |
54684461 | |
Doxycycline monohydrate |
Powder | |
167°C to 168°C | |
5 g | |
MFCD02682958 | |
Soluble in ethanol or methanol; Sparingly soluble in water;Soluble in dilute acids,alkali hydroxides and dimethyl sulfoxide. | |
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O | |
462.455 | |
462.46 (444.44 Anhydrous) |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P201-P202-P281-P308+P313-P501c
H361d
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only