missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 488
GREENER_CHOICE

Donkey Polyclonal Secondary Antibody

$308.81 - $723.96

Specifications

Antigen Rabbit IgG (H+L) Highly Cross-Adsorbed
Concentration 2 mg/mL
Content And Storage 4°C, store in dark
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIA32790
View Documents
Invitrogen™
A32790
1 mg N/A
Only null left
 
PIA32790TR
View Documents
Invitrogen™
A32790TR
100 ÎĽg N/A
Only null left
 
Description

Description

To minimize cross-reactivity, the donkey anti-Rabbit IgG whole antibodies have been cross-adsorbed against non-immunoglobulin rabbit serum proteins, IgG from bovine, goat, chicken, guinea pig, hamster, horse, sheep, mouse, rat, and human. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. Specificity: ...

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Rabbit IgG (H+L) Highly Cross-Adsorbed
4°C, store in dark
Polyclonal
Liquid
IgG
Rabbit
Gamma Immunoglobins Heavy and Light chains.
Secondary Antibody
2 mg/mL
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor Plus 488
Donkey
RUO
proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5
Secondary
Affinity chromatography
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'