missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecylbenzenesulfonic Acid (soft type) (mixture) 95.0+%, TCI America™

$41.02 - $80.16
Chemical Identifiers
CAS | 68584-22-5 |
---|---|
Molecular Formula | C18H30O3S |
Molecular Weight (g/mol) | 326.495 |
MDL Number | MFCD00066677 |
InChI Key | QJRVOJKLQNSNDB-UHFFFAOYSA-N |
Synonym | p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acidShow More |
PubChem CID | 29249 |
IUPAC Name | 4-dodecan-3-ylbenzenesulfonic acid |
SMILES | CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O |
Chemical Identifiers
68584-22-5 | |
326.495 | |
QJRVOJKLQNSNDB-UHFFFAOYSA-N | |
29249 | |
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O |
C18H30O3S | |
MFCD00066677 | |
p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acidShow More | |
4-dodecan-3-ylbenzenesulfonic acid |
Specifications
68584-22-5 | |
MFCD00066677 | |
2586 | |
QJRVOJKLQNSNDB-UHFFFAOYSA-N | |
4-dodecan-3-ylbenzenesulfonic acid | |
29249 | |
Liquid |
C18H30O3S | |
25 g | |
p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid | |
CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O | |
326.495 | |
≥95.0% (T) | |
Dodecylbenzenesulfonic Acid (soft type) (mixture) |
Safety and Handling
EINECSNumber : (3)-1884&(3)-1907
RTECSNumber : DB6600000
TSCA : Yes