Learn More
Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 255340050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dimethyl cis-1,2-cyclopropanedicarboxylate | |
1.1500g/mL | |
73°C | |
97% min. (GC) | |
C7H10O4 | |
MFCD00062805 | |
1.15 | |
Solubility in water: insoluble. Other solubilities: soluble in all organic solvents | |
COC(=O)C1CC1C(=O)OC | |
158.16 | |
158.16 |
826-34-6 | |
112.0°C (18.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4445 to 1.4465 | |
5g | |
dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-rel | |
JBVOSZYUSFDYIN-SYDPRGILSA-N | |
dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate | |
642979 | |
97+% |
Chemical Identifiers
826-34-6 | |
158.16 | |
JBVOSZYUSFDYIN-SYDPRGILSA-N | |
642979 | |
COC(=O)C1CC1C(=O)OC |
C7H10O4 | |
MFCD00062805 | |
dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-relShow More | |
dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
RUO â Research Use Only